To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:EAAT1/GLAST-1/SLC1A3
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 μl
- Environmentally Preferable:
- Gene ID:6507
- Antigen Synonyms:GLASTGLAST1|solute carrier family 1 (glial high affinity glutamate transporter)|Solute carrier family 1 member 3|EAAT1GLAST-1|member 3|EA6FLJ25094|Sodium-dependent glutamate/aspartate transporter 1|excitatory amino acid transporter 1
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [UniProt P24942]
- Purification:Immunogen affinity purified
- Cat. No.:103368-750
Specifications
About this item
The EAAT1 / GLAST-1 / SLC1A3 Antibody [PE] from Novus Biologicals is a rabbit polyclonal antibody to EAAT1 / GLAST-1 / SLC1A3. This antibody reacts with human, mouse, rat. The EAAT1 / GLAST-1 / SLC1A3 Antibody [PE] has been validated for the following applications: Flow Cytometry, Flow (Intracellular).
Type: Primary
Antigen: EAAT1/GLAST-1/SLC1A3
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat