To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:1810
- Antigen Synonyms:down-regulator of transcription 1|NC2|Negative cofactor 2-beta|protein Dr1|NC2-BETA|Down-regulator of transcription 1|TATA-binding protein-associated phosphoprotein|TBP-binding (negative cofactor 2)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:DR1 (NP_001929.1, 1 a.a. - 176 a.a.) full-length human protein. MASSSGNDDDLTIPRAAINKMIKETLPNVRVANDARELVVNCCTEFIHLISSEANEICNKSEKKTISPEHVIQALESLGFGSYISEVKEVLQECKTVALKRRKASSRLENLGIPEEELLRQQQELFAKARQQQAELAQQEWLQMQQAAQQAQLAAASASASNQAGSSQDEEDDDDI
- Purification:Protein A purified
- Cat. No.:103325-288
- Supplier no.:H00001810-B01P
Specifications
About this item
The DR1 Antibody from Novus Biologicals is a mouse polyclonal antibody to DR1. This antibody reacts with human, mouse, rat. The DR1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: DR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat