To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:DGCR8
- Clonality:Monoclonal
- Clone:4G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:54487
- Antigen Synonyms:DiGeorge syndrome critical region gene 8|C22orf12|DGCRK6chromosome 22 open reading frame 12|Gy1|microprocessor complex subunit DGCR8|DiGeorge syndrome critical region 8|pasha
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:DGCR8 (NP_073557, 675 a.a. ~ 772 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.LASQKILQLLHPHVKNWGSLLRMYGRESSKMVKQETSDKSVIELQQYAKKNKPNLHILSKLQEEMKRLAEEREETRKKPKMSIVASAQPGGEPLCTV
- Purification:IgG purified
- Cat. No.:103344-784
- Supplier no.:H00054487-M01
Specifications
About this item
The DGCR8 Antibody (4G3) from Novus Biologicals is a mouse monoclonal antibody to DGCR8. This antibody reacts with human. The DGCR8 Antibody (4G3) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DGCR8
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human