To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Dimethylarginine Dimethylaminohydrolase 1
- Antigen Symbol:DDAH1
- Clonality:Monoclonal
- Clone:3F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:23576
- Antigen Synonyms:DDAHdimethylargininase-1|dimethylarginine dimethylaminohydrolase 1FLJ25539|N(G)-dimethylarginine dimethylaminohydrolase 1|NG-dimethylarginine dimethylaminohydrolase|EC 3.5.3.18|DDAH-1|NG|Dimethylargininase-1|FLJ21264|DDAHI|N(G)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:DDAH1 (NP_036269, 181 a.a. ~ 286 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103341-548
- Supplier no.:H00023576-M01
Specifications
About this item
The Dimethylarginine Dimethylaminohydrolase 1 / DDAH1 Antibody (3F7) from Novus Biologicals is a mouse monoclonal antibody to Dimethylarginine Dimethylaminohydrolase 1 / DDAH1. This antibody reacts with human. The Dimethylarginine Dimethylaminohydrolase 1 / DDAH1 Antibody (3F7) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: DDAH1
Clonality: Monoclonal
Clone: 3F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human