To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:D4-GDI/RhoGDI2
- Clonality:Monoclonal
- Clone:10D774
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:397
- Antigen Synonyms:D4|LYGDI|RhoGDI2|Ly-GDI|GDIA2|RAP1GN1|GDID4|ARHGDIB Rho GDP dissociation inhibitor (GDI) beta
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen.
- Purification:Protein G purified
- Cat. No.:103360-226
Specifications
About this item
The D4-GDI / RhoGDI2 Antibody (10D774) [PE] from Novus Biologicals is a mouse monoclonal antibody to D4-GDI / RhoGDI2. This antibody reacts with human. The D4-GDI / RhoGDI2 Antibody (10D774) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: D4-GDI/RhoGDI2
Clonality: Monoclonal
Clone: 10D774
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human