To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CYFIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Epitope:internal region
- Gene ID:23191
- Antigen Synonyms:Sra-1|SHYC|KIAA0068cytoplasmic FMRP interacting protein 1|P140SRA-1|cytoplasmic FMR1 interacting protein 1|Specifically Rac1-associated protein 1|selective hybridizing clone|SRA1|cytoplasmic FMR1-interacting protein 1|FLJ45151
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Peptide with sequence C-TGSGRQEAQKTDAE corresponding to internal region according to NP_055423.1.
- Purification:Immunogen affinity purified
- Cat. No.:103405-988
The CYFIP1 Antibody from Novus Biologicals is a goat polyclonal antibody to CYFIP1. This antibody reacts with human. The CYFIP1 Antibody has been validated for the following applications: Peptide ELISA.
Type: Primary
Antigen: CYFIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal region
Host: Goat
Isotype: IgG
Reactivity: Human