To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CXCR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2833
- Antigen Synonyms:chemokine (C-X-C) receptor 3|C-X-C chemokine receptor type 3|CXC-R3|MigR|CD183|Interferon-inducible protein 10 receptor|CXCR-3|GPR9Mig-R|CD183 antigen|IP10 receptor|IP10-R|Mig receptor|chemokine (C-X-C motif) receptor 3|CMKAR3|CKR-L2IP-10 receptor|G protein-coupled receptor 9CD182
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CXCR3. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102707-160
- Supplier no.:NBP2-16044
Specifications
About this item
The CXCR3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CXCR3. This antibody reacts with human. The CXCR3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CXCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human