To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cullin 4a
- Antigen Symbol:CUL-4a
- Clonality:Monoclonal
- Clone:3H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:8451
- Antigen Synonyms:cullin 4A|cullin-4A|CUL-4A
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CUL4A (AAH08308, 1 a.a. - 100 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLYKQLRQACEDHVQAQILPFREDSLDSVLFLKKINACWQDHCRQMIMIRSIFLFLDRTYVLQNSTLPSIWDMGLELFRTHIISDKMVQSKTIDGILLLI
- Purification:IgG purified
- Cat. No.:103334-964
- Supplier no.:H00008451-M01
Specifications
About this item
The Cullin 4a Antibody (3H3) from Novus Biologicals is a mouse monoclonal antibody to Cullin 4a. This antibody reacts with human. The Cullin 4a Antibody (3H3) has been validated for the following applications: ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: CUL-4a
Clonality: Monoclonal
Clone: 3H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human