To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CNOT7
- Clonality:Monoclonal
- Clone:2C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:29883
- Antigen Synonyms:CAF-1|BTG1 binding factor 1|CCR4-NOT transcription complex subunit 7|hCAF-1|BTG1-binding factor 1|CCR4-NOT transcription complex|CAF1carbon catabolite repressor protein (CCR4)-associative factor 1|subunit 7|CCR4-associated factor 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CNOT7 (AAH60852, 1 a.a. ~ 286 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MPAATVDHSQRICEVWACNLDEEMKKIRQVIRKYNYVAMDTEFPGVVARPIGEFRSNADYQYQLLRCNVDLLKIIQLGLTFMNEQGEYPPGTSTWQFNFKFNLTEDMYAQDSIELLTTSGIQFKKHEEEGIETQYFAELLMTSGVVLCEGVKWLSFHSGYDFGYLIKILTNSNLPEEELDFFEILRLFFPVIYDVKYLMKSCKNLKGGLQEVAEQLELERIGPQHQAGSDSLLTGMAFFKMREMFFEDHIDDAKY
- Purification:IgG purified
- Cat. No.:103343-094
- Supplier no.:H00029883-M03
Specifications
About this item
The CNOT7 Antibody (2C4) from Novus Biologicals is a mouse monoclonal antibody to CNOT7. This antibody reacts with human. The CNOT7 Antibody (2C4) has been validated for the following applications: ELISA.
Type: Primary
Antigen: CNOT7
Clonality: Monoclonal
Clone: 2C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human