To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cytokeratin 7/13/17/18
- Antigen Symbol:CK7/13/17/18
- Clonality:Monoclonal
- Clone:LdS 23
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3855
- Antigen Synonyms:CK7|cytokeratin 7|Sarcolectin|SCLkeratin|type II cytoskeletal 7|K2C7|simple epithelial type I|type II mesothelial keratin K7|Cytokeratin-7|CK-7|Type-II keratin Kb7|keratin 7|K7keratin|K7|MGC3625|keratin-7|55K type II cytoskeletal|keratin|MGC129731
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Cytoskeletons of the human urothelial RT4 cell line
- Purification:Protein A or G purified
- Cat. No.:103357-604
Specifications
About this item
The Cytokeratin 7 / 13 / 17 / 18 Antibody (LdS 23) from Novus Biologicals is a mouse monoclonal antibody to Cytokeratin 7 / 13 / 17 / 18. This antibody reacts with human. The Cytokeratin 7 / 13 / 17 / 18 Antibody (LdS 23) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.
Type: Primary
Antigen: CK7/13/17/18
Clonality: Monoclonal
Clone: LdS 23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human