To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CHST9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:83539
- Antigen Synonyms:carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9|GALNAC4ST-2|GalNAc-4-ST2|FLJ42328|N-acetylgalactosamine 4-O-sulfotransferase 2|GalNAc-4-sulfotransferase 2|carbohydrate sulfotransferase 9|EC 2.8.2.-|N-acetylgalactosamine-4-O-sulfotransferase 2|GalNAc-4-O-sulfotransferase 2
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human CHST9. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102706-874
- Supplier no.:NBP2-15898
Specifications
About this item
The CHST9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CHST9. This antibody reacts with human, mouse. The CHST9 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: CHST9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse