To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Calponin 1
- Clonality:Monoclonal
- Clone:CNN1/832
- Conjugation:DyLight® 405
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1264
- Antigen Synonyms:smooth muscle|SMCCcalponin-1|calponins|Calponin H1|calponin 1|Basic calponin|Sm-Calp|basic
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human CNN1 protein
- Purification:Protein A or G purified
- Cat. No.:103395-542
Specifications
About this item
The Calponin 1 Antibody (CNN1 / 832) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to Calponin 1. This antibody reacts with human, rat. The Calponin 1 Antibody (CNN1 / 832) [DyLight 405] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Calponin 1
Clonality: Monoclonal
Clone: CNN1/832
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat