Order Entry
United States
ContactUsLinkComponent
Anti-Calbindin D-28K Chicken Polyclonal Antibody
Anti-Calbindin D-28K Chicken Polyclonal Antibody
Catalog # 103407-366
Supplier:  Novus Biologicals
Anti-Calbindin D-28K Chicken Polyclonal Antibody
Catalog # 103407-366
Supplier:  Novus Biologicals
Supplier Number:  NBP2-50028
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Calbindin D-28K
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Chicken
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgY
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    793
  • Antigen Synonyms:
    28kDa|RTVL-H protein|Calbindin D28|CALB|D-28K|calbindin|avian-type|CAB27|calbindin 1|Vitamin D-dependent calcium-binding protein|(28kD)
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    Full-length recombinant human Calbindin D-28K. [UniProt# P05937]
  • Purification:
    Ammonium sulfate precipitation
  • Cat. No.:
    103407-366

Specifications

About this item

The Calbindin D-28K Antibody from Novus Biologicals is a chicken polyclonal antibody to Calbindin D-28K. This antibody reacts with human, mouse, rat, bovine. The Calbindin D-28K Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Calbindin D-28K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat, Bovine