To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Calbindin D-28K
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoFluorescence:Yes
- Isotype:IgY
- Reactivity:Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:0.025ml
- Environmentally Preferable:
- Gene ID:793
- Antigen Synonyms:28kDa|RTVL-H protein|Calbindin D28|CALB|D-28K|calbindin|avian-type|CAB27|calbindin 1|Vitamin D-dependent calcium-binding protein|(28kD)
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full-length recombinant human Calbindin D-28K. [UniProt# P05937]
- Purification:Ammonium sulfate precipitation
- Cat. No.:103407-364
Specifications
About this item
The Calbindin D-28K Antibody from Novus Biologicals is a chicken polyclonal antibody to Calbindin D-28K. This antibody reacts with human, mouse, rat, bovine. The Calbindin D-28K Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Calbindin D-28K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat, Bovine