To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CCL7/MCP-3/MARC
- Clonality:Monoclonal
- Clone:4B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:6354
- Antigen Synonyms:chemokine (C-C motif) ligand 7|MCP3Monocyte chemotactic protein 3|MGC138465|small inducible cytokine A7 (monocyte chemotactic protein 3)|Monocyte chemoattractant protein 3|SCYA7MGC138463|MARC|MCP-3Small-inducible cytokine A7|C-C motif chemokine 7|NC28SCYA6|FIC
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CCL7 (NP_006264, 24 a.a. - 99 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QPVGINTSTTCCYRFINKKIPKQRLESYRRTTSSHCPREAVIFKTKLDKEICADPTQKWVQDFMKHLDKKTQTPKL
- Purification:IgG purified
- Cat. No.:103309-886
- Supplier no.:H00006354-M03
The CCL7 / MCP-3 / MARC Antibody (4B5) from Novus Biologicals is a mouse monoclonal antibody to CCL7 / MCP-3 / MARC. This antibody reacts with human. The CCL7 / MCP-3 / MARC Antibody (4B5) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CCL7/MCP-3/MARC
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human