To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CaMKIV
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:814
- Antigen Synonyms:calcium/calmodulin-dependent protein kinase type IV|CaMK-GR|CAM kinase- GR|CaMK IV|MGC36771|EC 2.7.11|calcium/calmodulin-dependent protein kinase IV|brain Ca++-calmodulin-dependent protein kinase type IV|brain Ca(2+)-calmodulin-dependent protein kinase type IV|CAM kinase IV|caMK|EC 2.7.11.17|calcium/calmodulin-dependent protein kinase type IV catalytic chain|IV|CaM kinase-GR
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:GNEDMKAIPEGEKIQGDGAQAAVKGAQAELMKVQALEKVKGADINAEEAPKMVPKAVEDGIKVADLELEEGLAEEKLKTVEEAAAPREGQGSSAVGFEVPQQ
- Purification:Immunogen affinity purified
- Cat. No.:103274-656
- Supplier no.:NBP1-88210
Specifications
About this item
The CaMKIV Antibody from Novus Biologicals is a rabbit polyclonal antibody to CaMKIV. This antibody reacts with human. The CaMKIV Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Type: Primary
Antigen: CaMKIV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human