To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:BMP-9
- Clonality:Monoclonal
- Clone:4D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2658
- Antigen Synonyms:BMP9BMP-9Bone morphogenetic protein 9|GDF-2|growth differentiation factor 2|growth/differentiation factor 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GDF2 (NP_057288 320 a.a. - 419 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SAGAGSHCQKTSLRVNFEDIGWDSWIIAPKEYEAYECKGGCFFPLADDVTPTKHAIVQTLVHLKFPTKVGKACCVPTKLSPISVLYKDDMGVPTLKYHYE
- Purification:IgG purified
- Cat. No.:103327-124
- Supplier no.:H00002658-M01
The BMP-9 Antibody (4D2) from Novus Biologicals is a mouse monoclonal antibody to BMP-9. This antibody reacts with human. The BMP-9 Antibody (4D2) has been validated for the following applications: ELISA.
Type: Primary
Antigen: BMP-9
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human