Order Entry
United States
ContactUsLinkComponent
Anti-Axl Mouse Monoclonal Antibody [clone: 7E10]
Anti-Axl Mouse Monoclonal Antibody [clone: 7E10]
Catalog # 103403-796
Supplier:  Novus Biologicals
Anti-Axl Mouse Monoclonal Antibody [clone: 7E10]
Catalog # 103403-796
Supplier:  Novus Biologicals
Supplier Number:  NBP2-37675
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    Axl
  • Clonality:
    Monoclonal
  • Clone:
    7E10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    558
  • Antigen Synonyms:
    AXL transforming sequence/gene|EC 2.7.10|JTK11|EC 2.7.10.1|UFOoncogene AXL|tyrosine-protein kinase receptor UFO|AXL oncogene|AXL receptor tyrosine kinase
  • Storage Buffer:
    Ascites
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    Purified recombinant extracellular fragment of human AXL fused with hIgGFc tag expressed in HEK293 cell line.
  • Purification:
    Unpurified
  • Cat. No.:
    103403-796

Specifications

About this item

The Axl Antibody (7E10) from Novus Biologicals is a mouse monoclonal antibody to Axl. This antibody reacts with human. The Axl Antibody (7E10) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Axl
Clonality: Monoclonal
Clone: 7E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human