To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Aminopeptidase PILS
- Antigen Symbol:ARTS1
- Clonality:Monoclonal
- Clone:4H8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:51752
- Antigen Synonyms:ERAAP1|endoplasmic reticulum aminopeptidase associated with antigen processing|ARTS-1PILSAP|EC 3.4.11.1|EC 3.4.11.-|endoplasmic reticulum aminopeptidase 1|Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator|Puromycin-insensitive leucyl-specific aminopeptidase|PILS-APERAAP|Aminopeptidase PILS|A-LAPALAP|EC 3.4.11|Adipocyte-derived leucine aminopeptidase|ARTS1aminopeptidase regulator of TNFR1 shedding|KIAA0525APPILS
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ARTS-1 (AAH30775, 832 a.a. ~ 942 a.a) partial recombinant protein with GST tag.FPQILTLIGRNPVGYPLAWQFLRKNWNKLVQKFELGSSSIAHMVMGTTNQFSTRTRLEEVKGFFSSLKENGSQLRCVQQTIETIEENIGWMDKNFDKIRVWLQSEKLERM
- Purification:IgG purified
- Cat. No.:103344-474
- Supplier no.:H00051752-M01
Specifications
About this item
The Aminopeptidase PILS / ARTS1 Antibody (4H8) from Novus Biologicals is a mouse monoclonal antibody to Aminopeptidase PILS / ARTS1. This antibody reacts with human. The Aminopeptidase PILS / ARTS1 Antibody (4H8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: ARTS1
Clonality: Monoclonal
Clone: 4H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human