To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein A5
- Antigen Symbol:APOAV
- Clonality:Monoclonal
- Clone:1G5G9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:116519
- Antigen Synonyms:apo-AV|MGC126836|FLJ97995|Apolipoprotein A5|Regeneration-associated protein 3|APOAV|APOA-V|apolipoprotein A-V|RAP3MGC126838
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788]
- Purification:Protein G purified
- Cat. No.:103405-212
Specifications
About this item
The Apolipoprotein A5 Antibody (1G5G9) [FITC] from Novus Biologicals is a mouse monoclonal antibody to Apolipoprotein A5. This antibody reacts with human. The Apolipoprotein A5 Antibody (1G5G9) [FITC] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: APOAV
Clonality: Monoclonal
Clone: 1G5G9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human