To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Angiomotin like 1
- Antigen Symbol:AMOTL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:154810
- Antigen Synonyms:junction-enriched and associated protein|JEAP|angiomotin like 1|angiomotin-like protein 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:AMOTL1 (NP_570899.1, 1 a.a. ~ 956 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103347-420
- Supplier no.:H00154810-D01P
The Angiomotin like 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Angiomotin like 1. This antibody reacts with human, mouse. The Angiomotin like 1 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: AMOTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse