To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:AG-2
- Clonality:Monoclonal
- Clone:1C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10551
- Antigen Synonyms:AG2member 17|secreted cement gland homolog|hAG-2|AG-2|anterior gradient 2 homolog (Xenopus laevis)|anterior gradient homolog 2 (Xenopus laevis)|HPC8|GOB-4
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:AGR2 (AAH15503, 1 a.a. ~ 176 a.a) full length recombinant protein with GST tag.MEKIPVSAFLLLVALSYTLARDTTVKPGAKKDTKDSRPKLPQTLSRGWGDQLIWTQTYEEALYKSKTSNKPLMIIHHLDECPHSQALKKVFAENKEIQKLAEQFVLLNLVYETTDKHLSPDGQYVPRIMFVDPSLTVRADITGRYSNRLYAYEPADTALLLDNMKKALKLLKTEL
- Purification:Protein A purified
- Cat. No.:103339-146
- Supplier no.:H00010551-M03
Specifications
About this item
The AG-2 Antibody (1C3) from Novus Biologicals is a mouse monoclonal antibody to AG-2. This antibody reacts with human. The AG-2 Antibody (1C3) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: AG-2
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human