To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Alcohol dehydrogenase 5
- Antigen Symbol:ADH5
- Clonality:Monoclonal
- Clone:12F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Cross Adsorption:No
- Gene ID:128
- Antigen Synonyms:FDHGSNOR|GSH-FDH|Alcohol dehydrogenase class chi chain|EC 1.1.1.-|ADHXEC 1.1.1.1|alcohol dehydrogenase (class III)|S-(hydroxymethyl)glutathione dehydrogenase|Glutathione-dependent formaldehyde dehydrogenase|formaldehyde dehydrogenase|FALDH|alcohol dehydrogenase 5 (class III)|ADH-3|EC 1.1.1|alcohol dehydrogenase class-3|Alcohol dehydrogenase 5|chi polypeptide|Alcohol dehydrogenase class-III|EC 1.1.1.284
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ADH5 (AAH14665, 1 a.a. - 374 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MANEVIKCKAAVAWEAGKPLSIEEIEVAPPKAHEVRIKIIATAVCHTDAYTLSGADPEGCFPVILGHEGAGIVESVGEGVTKLKAGDTVIPLYIPQCGECKFCLNPKTNLCQKIRVTQGKGLMPDGTSRFTCKGKTILHYMGTSTFSEYTVVADISVAKIDPLAPLDKVCLLGCGISTGYGAAVNTAKLEPGSVCAVFGLGGVGLAVIMGCKVAGASRIIGVDINKDKFARAKEFGATECINPQDFSKPIQEVLIEMTDGGVDYSFECIGNVKVMRAALEACHKGWGVSVVVGVAASGGEIATRPFQLVTGRTWKGTAFGGWKSVESVPKLVSEYMSKKIKVDEFVTHNLSFDEINKAFELMHSGKSIRTVVKI
- Purification:IgG purified
- Cat. No.:103321-444
- Supplier no.:H00000128-M01
Specifications
About this item
The alcohol dehydrogenase 5 Antibody (12F7) from Novus Biologicals is a mouse monoclonal antibody to alcohol dehydrogenase 5. This antibody reacts with human. The alcohol dehydrogenase 5 Antibody (12F7) has been validated for the following applications: Western Blot, ELISA.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: ADH5
Clonality: Monoclonal
Clone: 12F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human