To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ADAMTS17
- Clonality:Monoclonal
- Clone:3B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:170691
- Antigen Synonyms:EC 3.4.24.80|ADAMTS-17|FLJ16363|ADAM-TS17|EC 3.4.24.82|A disintegrin and metalloproteinase with thrombospondin motifs 17|17|a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif|ADAM metallopeptidase with thrombospondin type 1 motif|EC 3.4.24|EC 3.4.24.-|ADAM-TS 17|FLJ32769
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ADAMTS17 (NP_620688, 543 a.a. ~ 651 a.a) partial recombinant protein with GST tag.DGDWSPWGAWSMCSRTCGTGARFRQRKCDNPPPGPGGTHCPGASVEHAVCENLPCPKGLPSFRDQQCQAHDRLSPKKKGLLTAVVVDDKPCELYCSPLGKESPLLVAD
- Purification:IgG purified
- Cat. No.:103347-620
- Supplier no.:H00170691-M01
Specifications
About this item
The ADAMTS17 Antibody (3B7) from Novus Biologicals is a mouse monoclonal antibody to ADAMTS17. This antibody reacts with human, mouse. The ADAMTS17 Antibody (3B7) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ADAMTS17
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse