To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ABO, Blood Group A Antigen
- Antigen Symbol:ABO
- Clonality:Monoclonal
- Clone:3-3A
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:28
- Antigen Synonyms:A transferase|Histo-blood group A transferase|A3GALNT|Fucosylglycoprotein 3-alpha-galactosyltransferase|histo-blood group A2 transferase|Histo-blood group B transferase|NAGAT|Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase|EC 2.4.1.37|Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase|GTB|B transferase|histo-blood group ABO system transferase|A3GALT1|ABO blood group (transferase A|ABO glycosyltransferase|alpha 1-3-galactosyltransferase)|alpha 1-3-N-acetylgalactosaminyltransferase|Glycoprotein-fucosylgalactoside alpha-galactosyltransferase|transferase B|B(A) alpha-1,3-galactosyltransferase|EC 2.4.1.40
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Mucin isolated from an ovarian cyst fluid
- Purification:Protein A or G purified
- Cat. No.:103406-562
Specifications
About this item
The ABO, Blood Group A Antigen Antibody (3-3A) [FITC] from Novus Biologicals is a mouse monoclonal antibody to ABO, Blood Group A Antigen. This antibody reacts with human. The ABO, Blood Group A Antigen Antibody (3-3A) [FITC] has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ABO
Clonality: Monoclonal
Clone: 3-3A
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human