To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MARCH2
- Clonality:Monoclonal
- Clone:7F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:51257
- Antigen Synonyms:EC 6.3.2.-|MARCH-IIHSPC240|RNF172E3 ubiquitin-protein ligase MARCH2|Membrane-associated RING-CH protein II|Membrane-associated RING finger protein 2|RING finger protein 172|EC 6.3.2|membrane-associated ring finger (C3HC4) 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MARCH2 (NP_001005416, 1 a.a. ~ 176 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103343-898
- Supplier no.:H00051257-M04
Specifications
About this item
The MARCH2 Antibody (7F3) from Novus Biologicals is a mouse monoclonal antibody to 42796. This antibody reacts with human. The MARCH2 Antibody (7F3) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MARCH2
Clonality: Monoclonal
Clone: 7F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human