ContactUsLinkComponent
Somso® Intubation Manikin
Somso® Intubation Manikin
Catalog # 470233-264
Supplier:  MARCUS SOMMER
undefined
Somso® Intubation Manikin
Catalog # 470233-264
Supplier:  MARCUS SOMMER
Supplier Number:  CLA8
Specifications
  • Description:
    Intubation Model
  • Environmentally Preferable:
  • Cat. No.:
    470233-264
Specifications
About this item
An intubation model with feedback!
  • Can be combined with the CLA Nursing Doll
  • Provides alarm if to much pressure is exerted
  • No mechanics makes confirmation of success a stethoscope away
Natural size. This functional model, which has been developed in cooperation with the Federal Centre for Health Education in Cologne, makes it possible to learn how to intubate under life-like conditions. In its design and construction special importance has been placed on natural size and anatomical features to develop a functional model. If during intubation too much pressure is applied through the laryngoscope to the upper incisors, an acoustic signal is heard. The correct location of the tube, inserted after intubation, may be confirmed by audible respiratory sounds through a stethoscope lateral to the left and right wall of the thorax.