Human Exfoliated Deciduous Teeth Stem Cells (SHED), iXCells Biotechnologies
Catalog # 103521-928
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Size:0.5 million cells/vial
- Environmentally Preferable:
- Cat. No.:103521-928
Specifications
About this item
Human Exfoliated Deciduous Teeth Stem Cells (SHED) isolated from exfoliated deciduous teeth of young donors; Frozen
- High-quality Primary Human Exfoliated Deciduous Teeth Stem Cells (SHED)
- High purity
- IRB-approved
Negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast and fungi.