Order Entry
United States
ContactUsLinkComponent
SensoLyte® Thiol Quantitation Assay Kit (Colorimetric), AnaSpec
SensoLyte® Thiol Quantitation Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-476
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® Thiol Quantitation Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-476
Supplier:  Anaspec Inc
Supplier Number:  AS-72136
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    500 Assays
  • Environmentally Preferable:
  • Cat. No.:
    103010-476
  • Storage temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72136

Specifications

About this item

Thiols, compounds that contain sulfhydryl group, are powerful reducing agents capable of acting as antioxidants, enzyme cofactors, and neuromodulators.

The SensoLyte® Thiol Quantitation Assay Kit utilizes the widely used Elmann’s reagent for colorimetric measurement of thiol concentrations. The sulfhydryl group of thiol reacts with Ellman’s Reagent [5, 5’-dithiobis (2-nitrobenzoic acid)] and generates 2-nitro-5-thiobenzoic acid (TNB) producing a yellow color that can be detected at absorbance 420 nm. The intensity of the color produced is proportional to thiol concentration.