To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Beta-1,3-glucuronyltransferase 1
- Antigen Symbol:B3GAT1
- Clonality:Monoclonal
- Clone:TBO1
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Size:50 μg
- Environmentally Preferable:
- Format:SAFIRE Purified
- Form:liquid
- Antigen Synonyms:LEU7|GLCATP|CD57|GLCUATP|NK-1|NK|HNK1
- Cat. No.:75842-022
- Supplier no.:09611-25-50
Specifications
About this item
The TBO1 monoclonal antibody specifically binds to human CD57, a 110 kDA glycoprotein expressed on a subset of natural killer lymphocytes, cells, neural cells and striated muscle. It is reported that the molecule is involved in cell-matrix interactions and is upregulated in some disease states.
Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: TBO1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human