To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Assay Type:Sandwich
- Host:
- Primary antibody reactivity:Human
- Target protein:Lipocalin-2/NGAL
- Description:Human Lipocalin-2/NGAL EZ-Set; ELISA Kit (DIY Antibody Pairs)
- Size:For 5 plates, 96 wells each
- Environmentally Preferable:
- Cat. No.:76172-834
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Lipocalin-2/NGAL in Human samples
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Lipocalin-2/NGAL in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.
The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.