ContactUsLinkComponent
Fermantas DNA Gel Loading Dye, 6X, Thermo Scientific
Fermantas DNA Gel Loading Dye, 6X, Thermo Scientific
Catalog # 101672-984
Supplier:  Fermentas
CAS Number:  
undefined
Fermantas DNA Gel Loading Dye, 6X, Thermo Scientific
Catalog # 101672-984
Supplier:  Fermentas
Supplier Number:  R0611
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    DNA loading dye 6X
  • Size:
    5 x 1 mL
  • Environmentally Preferable:
  • Cat. No.:
    101672-984
  • Supplier no.:
    R0611
Specifications
About this item
This DNA loading dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels
  • No DNA masking during gel exposure to UV light
  • EDTA binds divalent metal ions and inhibits metal dependent nucleases
Ideal for preparation of DNA ladders, markers, and samples for loading on agarose or polyacrylamide gels.
It contains two different dyes (bromophenol blue and xylene cyanol FF) for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well.
Caution: For research use only. Not for use in diagnostic procedures.