Order Entry
United States
ContactUsLinkComponent
 
Novagen® Casein, alkali-soluble
  EM70955-3
 :  
Novagen® Casein, alkali-soluble
  EM70955-3
 :  70955-3
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Pack type:
    Glass bottle
  • Source:
  • Species:
  • Size:
    225 ml
  • Environmentally Preferable:
  • Storage Conditions:
    2 – 8 °C
  • Protein/Peptide Name:
    Casein, alkali-soluble
  • Cat. No.:
    EM70955-3
  • Supplier no.:
    70955-3

 

 

Alkali-Soluble Casein is a superior blocking reagent for Western blot applications.

This preparation minimizes non-specific background relative to other blocking reagents, such as BSA, gelatin, or non-fat milk. It is effective in colorimetric and chemiluminescent detection protocols. Note that due to its high coating properties, detection sensitivity for some antibodies may be lower than with other blocking reagents under the same conditions (although the background is also reduced); the concentration can be optimized for maximal signal:noise ratios. Recommended working concentration range is 0.25 – 1%.