5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] dye, Pierce™
Catalog # PI46112
Supplier: Avantor
CAS Number:
Specifications
- Size:10 mg
- Environmentally Preferable:
- Cat. No.:PI46112
- Supplier no.:46112
Specifications
Specification Test Results
- Absorption Spectrum(in MeOH):543 nm max.; E = 85000 cm-1 M-1
- Fluorescence Spectrum (in MeOH):EMmax = 572 nm.
- Basis for Identification:NMR is consistent with structure.
- Purity:87 % by HPLC at 254 nm
Specification Test Results
About this item
Synonyms: Tetramethylrhodamine-5(6)-isothiocyanate, 5(6)-TRITC, MRITC, TRITC
:
:443.52 g/mol
:GHS07
:GHS08
Storage Temperature:Freezer
CAS Number:95197-95-8
Formula:C₂₅H₂₁N₃O₃S
MDL Number:MFCD00063517