Hitachi M4 SEM Mount, Electron Microscopy Sciences
Hitachi M4 SEM mounts are made from lathe finished ultra-pure aluminum for scanning electron microscopy.
Surface: 1" (25 mm)
Height: 5/8" (16 mm)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.