About this item
The OneStep™ PCR Inhibitor Removal Kit allows for the efficient removal of contaminants that can inhibit downstream enzymatic reactions from DNA and RNA preparations.
- Removes PCR inhibitors such as polyphenolics, humic/fulvic acids, tannins, melanin, etc. from nucleic acid solutions to yield high quality DNA or RNA
- Fast, one-step procedure for cleaning impure samples prior to PCR, sequencing, reverse transcription (RT), etc
About OneStep™ PCR Inhibitor Removal Kit
The OneStep™ PCR Inhibitor Removal Kit contains all the components needed for efficient removal of contaminants that can inhibit downstream enzymatic reactions (e.g. PCR and RT) from DNA and RNA preparations. The column matrices have been specifically designed for the efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from the most impure DNA and RNA preparations. Sample clean-up is as simple as applying, spinning, and recovering a sample from the column.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: