Order Entry
United States
ContactUsLinkComponent
Mouse;Rat Kisspeptin-10 (Kp-10)
Mouse;Rat Kisspeptin-10 (Kp-10)
Catalog # 103008-422
Supplier:  Anaspec Inc
CAS Number:  
Mouse;Rat Kisspeptin-10 (Kp-10)
Catalog # 103008-422
Supplier:  Anaspec Inc
Supplier Number:  AS-64684
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Source:
  • Species:
    Rat
    Mouse
  • Size:
    1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    C-terminal Metastin Peptide
  • Protein/Peptide Name:
    Kisspeptin-10 (Kp-10)
  • Purity:
    95%
  • Molecular Weight:
    1318.5 Da
  • Sequence:
    YNWNSFGLRY-NH2
  • Cat. No.:
    103008-422
  • Supplier no.:
    AS-64684

Specifications

About this item

Kisspeptin was originally identified as a human metastasis suppressor gene that has the ability to suppress melanoma and breast cancer metastasis. Kisspeptin-GPR54 signaling has an important role in initiating secretion of gonadotropin-releasing hormone (GnRH) at puberty from the anterior pituitary.
This amidated peptide sequence is found in C-terminal residues 110 to 119 of the neurohormone Metastin (also referred to as Kisspeptin-10); it increases plasma concentrations of GH (Growth Hormone) and LH (Luteinizing Hormone).
Sequence:YNWNSFGLRY-NH2
MW:1318.5 Da
% peak area by HPLC:95
Storage condition:-20° C