Order Entry
United States
ContactUsLinkComponent
Colivelin
Colivelin
Catalog # 103007-098
Supplier:  Anaspec Inc
CAS Number:  
Colivelin
Catalog # 103007-098
Supplier:  Anaspec Inc
Supplier Number:  AS-61930
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Source:
  • Species:
  • Size:
    1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Activity-Dependent Neurotrophic Factor (ADNF) C-AGA-(C8R) HNG1 Peptide|hybrid colivelin Peptide|ADNF-HNG1 Peptide|hybrid Peptide
  • Protein/Peptide Name:
    Colivelin
  • Purity:
    95%
  • Molecular Weight:
    2645.1 Da
  • Sequence:
    SALLRSIPAPAGASRLLLLTGEIDLP
  • Cat. No.:
    103007-098
  • Supplier no.:
    AS-61930

Specifications

About this item

This hybrid peptide named colivelin was synthesized to potentiate the neuroprotective effect of humanin (HN). Colivelin is composed of Activity-Dependent Neurotrophic Factor (ADNF) C-terminally fused to AGA-(C8R) HNG17, a potent HN derivative. Colivelin completely suppresses cell death induced by overexpressed Familial Alzheimer's disease (FAD)-causative genes and beta-amyloid (1-43). Intraperitoneally administered colivelin suppresses memory impairment and might serve as a novel drug candidate for treatment of Alzheimer's disease.
Sequence:SALLRSIPAPAGASRLLLLTGEIDLP
MW:2645.1 Da
% peak area by HPLC:95
Storage condition:-20° C