Order Entry
United States
ContactUsLinkComponent
Chicken OVA (323-339),Biotin
Chicken OVA (323-339),Biotin
Catalog # 103008-440
Supplier:  Anaspec Inc
CAS Number:  
Chicken OVA (323-339),Biotin
Catalog # 103008-440
Supplier:  Anaspec Inc
Supplier Number:  AS-64767
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Biotin
  • Source:
  • Species:
    Chicken
  • Size:
    1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    OVA 323-339 biotin|OVA Peptide biotin
  • Protein/Peptide Name:
    OVA (323-339)
  • Purity:
    95%
  • Molecular Weight:
    2000.2 Da
  • Sequence:
    Biotin-ISQAVHAAHAEINEAGR
  • Cat. No.:
    103008-440
  • Supplier no.:
    AS-64767

Specifications

About this item

This is an N-terminal biotin-labeled OVA Peptide, amino acids 323 to 339. This peptide is an H-2b-restricted OVA class II epitope.
Sequence: Biotin-ISQAVHAAHAEINEAGR
MW: 2000.2 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C