To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Source:
- Species:HIV
- Size:1 mg
- Environmentally Preferable:
- Storage Conditions:–20 °C
- Protein Synonyms:C-TAT 48-57|Tat-C 48-57
- Protein/Peptide Name:Tat-C (48-57) Peptide
- Purity:95%
- Molecular Weight:1499.8 Da
- Sequence:CGRKKRRQRRR
- Cat. No.:103007-166
- Supplier no.:AS-62063
Specifications
About this item
This peptide is amino acids 48 to 57 fragment of TAT with an additional cysteine residue at the N-terminus. This peptide contains the protein transduction domain (PTD) of the HIV Tat protein that inhibits HSV-1 entry. The addition of a cysteine residue to the N-terminus of the Tat-PTD (Tat-C peptide) improves the antiviral activity against HSV-1 and HSV-2. Tat-C acts extracellularly, blocking entry of adsorbed virus immediately without eluting virions.
Sequence: CGRKKRRQRRR
MW: 1499.8 Da
% Peak area by HPLC: 95
Storage condition: -20°C