Order Entry
United States
ContactUsLinkComponent
Serum Free Cryo Medium, Quality Biological
Serum Free Cryo Medium, Quality Biological
Catalog # 76169-066
   Sustainable Packaging
CAS Number:  
undefined
Serum Free Cryo Medium, Quality Biological
Catalog # 76169-066
Supplier Number:  163-320-051
   Sustainable Packaging
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Pack type:
    Each (100 ml)
  • Description:
    Serum free cryo medium
  • Size:
    100 ml
  • Environmentally Preferable:
    Sustainable Packaging
  • Sterility:
    Sterile filtered
  • Environmental Certification:
    ECO LOGOS
  • Shipping Conditions:
    on dry ice
  • Cat. No.:
    76169-066
  • Shelf life:
    24 months
  • Application:
    A major component for the cryopreservation of cells grown under serum free conditions

Specifications

About this item

A sterile, 2X concentrated freezing medium containing DMSO developed for the cryopreservation of cells cultured in serum free conditions.

  • Animal-free, protein-free, chemically defined cryopreservation medium Without phenol red
  • Contains DMSO and other cryoprotective components
  • 2X concentration
  • Absence of FBS allows continuous culture and preservation of cells under serum free condition

Serum Free Cryo Medium's 2X concentrated formulation allows for dilution with either conditioned medium or complete fresh medium. Serum Free Cryo Medium can be easily paired with serum free medium, QBSF-55 ( #10128-230) for culturing cells in serum free conditions.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: Accesory not included.
Caution: Research Use Only