Specifications
- Description:Antiseptic Spray, with Pump
- Size:4 oz.
- Environmentally Preferable:
- Cat. No.:76048-936
Specifications
About this item
First aid antiseptic pump spray decreases germs in minor cuts and scrapes.
- Decreases germs in minor cuts and scrapes
- Ideal for infection prevention, skin abrasions, and cuts
It is quick and easy to use: Spray a small amount of the product on the affected area 1 to 3 times daily. May be covered with a sterile bandage. If bandaged, allow to dry first. The antiseptic spray is ideal for infection prevention, skin abrasions, and cuts.