Blood Filter Strips, Advantec
Catalog # 101104-522
Supplier: Advantec MFS
Specifications
- Description:Blood Filter Strips
- Environmentally Preferable:
- Cat. No.:101104-522
- Supplier no.:800700
Specifications
About this item
These blood filter strips are used to track healthy and diseased animal populations safely, efficiently, and economically.
- High purity cellulose fibers for consistent, uniform sampling
- Each strip holds approximately 1 ml of blood or 0.04 ml serum
- Filter strips require no refrigeration or pretreatment for sampling, storage, and analysis