Flame Resistant/ARC Bib Style Rain Pants, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Bib Style Rain Pant for visibility and protection from the elements, as well as flash fire and electrical arc flash.
- 10 oz. polyurethane over FR cotton knit
- 16" zippered boot openings
- Non-metallic quick release suspender clips
- Cargo pocket
Lakeland's Flame Resistant/ARC Rain Bibs are designed to protect wearers from the elements as well as electrical arc and flash fire hazards, all while maintaining visibility on the job. Features include cargo pocket, 2-snap fly and non-metallic, quick release suspender clips. The 16" leg zippers make for easy donning and doffing, even over the largest work boots.
Certifications: ANSI 107-2010 Class E. ASTM F1891. ASTM F2733.
Specifications
- Catalog No:
- 75923-738
- 75923-606
- 75923-608
- 75923-610
- 75923-596
- 75923-598
- 75923-600
- 75923-602
- 75923-604
- Garment application:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Weight:
- 10 oz.
- 10 oz.
- 10 oz.
- 10 oz.
- 10 oz.
- 10 oz.
- 10 oz.
- 10 oz.
- 10 oz.
Specifications
Frequently Bought Together