Order Entry
United States
ContactUsLinkComponent
OptiPlate-384, White Opaque 384-well Microplate, PerkinElmer
OptiPlate-384, White Opaque 384-well Microplate, PerkinElmer
Catalog # 101679-990
Supplier:  Avantor
undefined
OptiPlate-384, White Opaque 384-well Microplate, PerkinElmer
Catalog # 101679-990
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:
  • Detection Method:
    Time-Resolved Fluorescence (TRF),Luminescence,Alpha,Radiometric
  • Shipping Conditions:
    Shipped Ambient
  • Cat. No.:
    101679-990
  • Storage temperature:
    19...25 °C
  • Supplier no.:
    6007290

Specifications

About this item

The OptiPlate is one of the most versatile Microplates offered by PerkinElmer, used in many different applications, assays, and instruments

  • Harmonized Code US-3926909910
  • Material-Polystyrene

The optimized well design and high quality plastics provides low background and crosstalk. >OptiPlates are uniquely engineered to be the same height for 96-, 384-, and 1536-well plates, and with extraordinary well flatness for ease in automation. Leveraging years of assay and instrument experience in plate detection, PerkinElmer designs better Microplates for better performance that guarantees better results for all PerkinElmer applications.