Westex DH Flame Resistant Coverall, Navy Blue, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Inherently flame resistant coveralls for primary protection from flash fire and electrical arc flash.
- Inherently flame resistant fabric will not lose flame resistant properties
- Pleated back and elastic waist for fit and comfort
- Flared leg cuffs for ease of donning and doffing
Westex DH Flame Resistant Coveralls are dual certified for both flash fire and electrical arc flash protection. Designed for fit and comfort with pleated back, elastic waist, and plenty of pockets. Flared leg openings make donning and doffing easier and sleeve cuffs feature adjustable snap closures.
Certifications: NFPA 2112, UL certified for flash fire protection. NFPA 70E, CAT2 electrical arc flash protection.
Specifications
- Catalog No:
- 75923-444
- 75923-442
- 75923-440
- 75923-438
- 75923-436
- 75927-796
- 75927-798
- 75927-800
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Risk area:
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening