To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Source:
- Species:
- Size:250 units
- Environmentally Preferable:
- Enzyme Name:RNase R
- Concentration:20 units/μl
- Cat. No.:76081-690
A 3´ to 5´ exoribonuclease that digests all linear RNAs except lariat or circular RNA structures making it well-suited to RNA splicing and other research applications
- Specific: Digests all linear RNAs but does not digest lariat or circular RNA structures, or doublestranded RNA with 3' overhangs shorter than seven nucleotides
- Heat Inactivated: Heat at 65°C for 20 minutes to kill enzyme activity
- Valuable: Use the unique properties of this exoribonuclease to study alternative splicing and gene expression
Ribonuclease R (RNase R) from E
Lariat RNAs isolated using this method can be used as a template to produce labeled cDNA as a target for microarrays containing potential intron sequences, or for tiling arrays containing overlapping regions of complete chromosomes or genomes. The cDNA produced will not be a linear representation of the intron, but the sequences contained in it will be intron-derived.
Applications include:
Alternative splicing and gene expression studies
Intron cDNA production
Intronic screening of cDNA libraries
coli is a magnesium-dependent 3´ to 5´ exoribonuclease that digests essentially all linear RNAs but does not digest lariat or circular RNA structures, or doublestranded RNA with 3´ overhangs shorter than seven nucleotides. Most cellular RNAs will be digested completely by RNase R, with the exception of tRNAs, 5S RNA, and intron lariats. The 3´ tails of lariats will be trimmed by RNase R to the branch point nucleotide, where there is a 2´,5´-phosphodiester linkage. Lariats are produced during pre-mRNA splicing of intron regions and can be isolated from a mixture of total RNA by digestion with RNase R.