Order Entry
United States
ContactUsLinkComponent
LaMotte® Aquaponics Test Kit
LaMotte® Aquaponics Test Kit
Catalog # 470228-152
Supplier:  LaMotte
CAS Number:  
undefined
LaMotte® Aquaponics Test Kit
Catalog # 470228-152
Supplier:  LaMotte
Supplier Number:  3637
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Aquaponics Test Kit
  • Environmentally Preferable:
  • Cat. No.:
    470228-152
  • Range:
    Ammonia 0-2 ppm;
    Nitrite 0.05-0.8 ppm;
    Nitrate 0.25-10.0 ppm;
    pH 5.0-10.0 pH;
    Iron, Ferrous and Ferric 0.5-10.0 ppm;
    Alkalinity 0-200 ppm;
    Dissolved Oxygen 0-10 ppm
  • No. of tests:
    50

Specifications

About this item

Aquaponics is growing rapidly as a hobby and as an industry as the farm-to-table trend continues to spread.

  • Includes materials to perform 50 tests
  • Detailed instruction manual

Designed for small to midsize systems, this test kit monitors basic water quality for both the aquaculture and hydroponics segments of your system. A detailed, diagrammed instruction manual is provided along with a quick reference lid instruction. Reagents, labware and accessories are mounted in a foam-lined carrying case. Kit is complete with labware, accessories, and reagents to perform approximately 50 repetitions per test factor. Test chemistries and comparators are compatible with fresh, brackish, or salt water systems.