About this item
Homogenous, TR-FRET based assay for detection of cAMP
- Homogenous assay , mix and read
- No wash steps
- Eliminates compound interference
- Allows for HTS assays to be performed in a miniaturized format.
- TR-FRET format increases sensitivity
This kit is designed to to detect and quantitatively measure the amount of cyclic AMP (cAMP) present in cell based assay systems. A europium (Eu) labeled monoclonal anti-cAMP antibody (Donor) is paired with a fluorescently labeled cAMP analog (Acceptor), and in the absence of any endogenous cAMP, will produce a time-resolved energy transfer signal at 665 nm when excited at 340 nm. If cAMP accumulates in the well due to the activation of cellular adenylyl cyclase, this cAMP inhibits the formation of the labeled Donor/Acceptor complex thereby decreasing the amount of energy transfer signal produced and the overall drop in signal is inversely proportional to the amount of cAMP accumulated.