InVivoMab anti-human CD32 (FcγRIIA), Clone: IV.3, Isotype: mouse IgG2b, Applications: in vivo FcγRIIA blockade in humanized mice, in vitro FcγRIIA blockade, ELISA, Flow cytometry
Type: Primary
Antigen: CD32 (FcgammaRIIA)
Clonality: Monoclonal
Clone: IV.3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76556-304
- 76556-184
- 76556-078
- 76557-288
- 76556-438
- Antigen Symbol:
- CD32 (FcgammaRIIA)
- CD32 (FcgammaRIIA)
- CD32
- CD32 (FcgammaRIIA)
- CD32 (FcgammaRIIA)
- Antigen Name:
- CD32 (FcgammaRIIA)
- CD32 (FcgammaRIIA)
- CD32 (FcgammaRIIA)
- CD32 (FcgammaRIIA)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- IV.3
- IV.3
- IV.3
- IV.3
- IV.3
- Reactivity:
- Human
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- ELISA:
- Yes
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes